Sodium tripolyphosphate
Sodium tripolyphosphate
Sodium tripolyphosphate
Sodium tripolyphosphate
Thermo Scientific Chemicals
Sodium tripolyphosphate
CAS: 7758-29-4 | Na5O10P3 | 367.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF013440.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium tripolyphosphate
Melting Point622°C
CAS7758-29-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
View more
Sodium tripolyphosphate is used in water softening, heavy-duty fabric washing, automatic dish washing, drilling fluid cleaning and liquid cattle feed supplements. It is also used as a peptizing, emulsifying, dispersing agent and as a hydrogen peroxide solution stabilizer. It acts as a preservative for seafood, meats, poultry, and animal feeds. It finds application as a polyanion crosslinker in polysaccharide based drug delivery. Further, it is used as a sealant for leaking farm ponds and a calcium and magnesium chelating agent. In addition to this, it is used in ceramics, leather tanning, flame retardants, paper pulping, paper coatings, textile processing, ore flotation, anticorrosion pigments and rubber production.