Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min
Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min
Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min
Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min
Thermo Scientific Chemicals
Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min
CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF013437.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydrogen phosphate, anhydrous
CAS7558-79-4
ColorWhite
Health Hazard 1H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
View more
Sodium hydrogen phosphate is used in food and water treatment in conjunction with trisodium phosphate. It is used as an emulsifier and buffer in foods and pharmaceuticals, anti-caking additive in powdered products, dietary supplement, scale inhibitor, reagent and buffer in analytical chemistry. It finds application in detergents, cleaning agents, textile, leather auxiliary, fertilizers and galvanoplastics. Further, it is used as a saline laxative to treat constipation. It inhibits the calcium scale formation in water treatment plants.