Check Order Status
H-Aib-OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
H-Aib-OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013407.36
100 g
Catalog number
ALF013407.36
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
H-Aib-OH
CAS
62-57-7
Color
White/Off-White
Molecular Formula
C4H9NO2
InChI Key
FUOOLUPWFVMBKG-UHFFFAOYSA-N
View more