Check Order Status
Fmoc-D-Bip-OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Fmoc-D-Bip-OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013407.14
5 g
Catalog number
ALF013407.14
Price (MYR)
241.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
241.00
EA
Add to cart
Specifications
Chemical Name or Material
Fmoc-D-Bip-OH
CAS
205526-38-1
Color
White/Off-White
Molecular Formula
C30H25NO4
InChI Key
VSGACONKQRJFGX-UHFFFAOYNA-N
View more