Check Order Status
Fmoc-Cys p-MeO-Bzl -OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Fmoc-Cys p-MeO-Bzl -OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013406.30
5 g
Catalog number
ALF013406.30
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
Fmoc-Cys p-MeO-Bzl -OH
CAS
141892-41-3
Color
White/Off-White
Molecular Formula
C26H25NO5S
InChI Key
IWZGYHFOLFRYPK-UHFFFAOYNA-N
View more