Check Order Status
Fmoc-Cys Bzl -OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Fmoc-Cys Bzl -OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013406.18
1 g
Catalog number
ALF013406.18
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material
Fmoc-Cys Bzl -OH
Color
White/Off-White
Quantity
1 g
Grade
Biochemical