Check Order Status
Fmoc-Glu OBut -OH, Advanced ChemTech
Element Search
Structure Search
Thermo Scientific Chemicals
Fmoc-Glu OBut -OH, Advanced ChemTech
Have Questions?
Catalog Number
Quantity
ALF013405.30
25 g
Catalog number
ALF013405.30
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material
Fmoc-Glu OBut -OH
CAS
71989-18-9
Color
White to Off-White
Molecular Formula
C24H27NO6
InChI Key
GOPWHXPXSPIIQZ-UHFFFAOYNA-N
View more