Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
Thermo Scientific Chemicals
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)
CAS: 14637-88-8 | C15H21DyO6 | 459.83 g/mol
Have Questions?
Catalog NumberQuantity
ALF013219.1425 g
Catalog number ALF013219.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDysprosium(III) 2,4-pentanedionate hydrate
CAS14637-88-8
Recommended StorageAmbient temperatures
Physical FormPowder
Quantity25 g