Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)
CAS: 181266-82-0 | C15H21EuO6 | 449.29 g/mol
Have Questions?
Catalog NumberQuantity
ALF013209.0910 g
Catalog number ALF013209.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEuropium(III) 2,4-pentanedionate hydrate
CAS181266-82-0
Melting Point140°C (decomposition)
Recommended StorageAmbient temperatures
EINECS Number238-185-7
View more
It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.