Ammonium bromide, Puratronic™, 99.999% (metals basis)
Ammonium bromide, Puratronic™, 99.999% (metals basis)
Ammonium bromide, Puratronic™, 99.999% (metals basis)
Ammonium bromide, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals
Ammonium bromide, Puratronic™, 99.999% (metals basis)
CAS: 12124-97-9 | BrH4N | 97.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF012981.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium bromide
Melting Point452°C (sublimation)
CAS12124-97-9
ColorWhite
Health Hazard 1H319
View more
Ammonium bromide is used in pharmaceuticals for its bromide ion. It is used for a variety of analytical chemistry applications. It is also used for photography in films, plates and papers; in fireproofing of wood; in lithography and process engraving and as corrosion inhibitors.