Dysprosium(III) nitrate pentahydrate, 99.9% (REO)
Dysprosium(III) nitrate pentahydrate, 99.9% (REO)
Dysprosium(III) nitrate pentahydrate, 99.9% (REO)
Dysprosium(III) nitrate pentahydrate, 99.9% (REO)
Dysprosium(III) nitrate pentahydrate, 99.9% (REO)
Thermo Scientific Chemicals
Dysprosium(III) nitrate pentahydrate, 99.9% (REO)
CAS: 10031-49-9 | DyH10N3O14 | 438.59 g/mol
Have Questions?
Catalog NumberQuantity
ALF012922.22100 g
Catalog number ALF012922.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDysprosium(III) nitrate pentahydrate
Melting Point88.6°C
Name NoteRare Earth Oxides Content
CAS10031-49-9
ColorYellow
View more
Dysprosium(III) nitrate pentahydrate is used to prepare carboxylate-based dinuclear dysprosium compound by reacting with n-butyric acid and triethylamine using methanol as a solvent.