Erbium(III) nitrate hydrate, 99.9% (REO)
Erbium(III) nitrate hydrate, 99.9% (REO)
Erbium(III) nitrate hydrate, 99.9% (REO)
Erbium(III) nitrate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Erbium(III) nitrate hydrate, 99.9% (REO)
CAS: 10031-51-3 | ErH10N3O14 | 443.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF012920.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialErbium(III) nitrate hydrate
Melting Point130°C (in -4H2O)
CAS10031-51-3
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Used as laboratory chemicals.