Neodymium(III) nitrate hexahydrate, 99.9% (REO)
Neodymium(III) nitrate hexahydrate, 99.9% (REO)
Neodymium(III) nitrate hexahydrate, 99.9% (REO)
Neodymium(III) nitrate hexahydrate, 99.9% (REO)
Thermo Scientific Chemicals
Neodymium(III) nitrate hexahydrate, 99.9% (REO)
CAS: 16454-60-7 | H12N3NdO15 | 438.344 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF012912.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNeodymium(III) nitrate hexahydrate
Melting Point∼40°C
CAS16454-60-7
Health Hazard 1H272
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Neodymium(III) nitrate hexahydrate is used to prepare nanoparticles of neodymium oxybromide and neodymium oxide. It is also used in coloring glasses and for doping some glass lasers.