Praseodymium(III) nitrate hydrate, 99.9% (REO)
Praseodymium(III) nitrate hydrate, 99.9% (REO)
Praseodymium(III) nitrate hydrate, 99.9% (REO)
Praseodymium(III) nitrate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Praseodymium(III) nitrate hydrate, 99.9% (REO)
CAS: 15878-77-0 | H12N3O15Pr | 435.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF012909.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPraseodymium(III) nitrate hydrate
Name NoteRare Earth Oxides Content
CAS15878-77-0
ColorGreen
Health Hazard 1H272-H315-H319-H335
View more
Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.