Samarium(III) nitrate hexahydrate, 99.9% (REO)
Samarium(III) nitrate hexahydrate, 99.9% (REO)
Samarium(III) nitrate hexahydrate, 99.9% (REO)
Samarium(III) nitrate hexahydrate, 99.9% (REO)
Thermo Scientific Chemicals
Samarium(III) nitrate hexahydrate, 99.9% (REO)
CAS: 13759-83-6 | H12N3O15Sm | 444.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF012906.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSamarium(III) nitrate hexahydrate
Melting Point78°C to 79°C
CAS13759-83-6
Health Hazard 1H272-H315-H320-H335
Health Hazard 3P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c
View more
It is used in NMR spectroscopy for spectra resolution and is used for doping purpose. It is also used as a chemical reagent.