Ytterbium(III) nitrate hydrate, 99.9% (REO)
Ytterbium(III) nitrate hydrate, 99.9% (REO)
Thermo Scientific Chemicals
Ytterbium(III) nitrate hydrate, 99.9% (REO)
Have Questions?
Catalog NumberQuantity
ALF012901.1850 g
Catalog number ALF012901.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialYtterbium(III) nitrate hydrate
ColorWhite
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Ytterbium(III) nitrate hydrate is used as a precursor of nanoscale coatings of carbon composites. The ytterbium +3 ion is used as a doping material in solid state lasers and double clad fiber lasers. It is also used to make yttrium-based surfactant mesophases, which has adsorbing as well as optically functional properties.