Platinum black, HiSPEC™ 1000
Platinum black, HiSPEC™ 1000
Platinum black, HiSPEC™ 1000
Platinum black, HiSPEC™ 1000
Platinum black, HiSPEC™ 1000
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
0.25 g
1 g
Catalog number ALF012755.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum black
CAS7440-06-4
Health Hazard 1H228: Flammable solid.
AppearanceDark gray powder
Density21.45g/cm3 at 20°C
View more
Platinum black is most commonly used for electrolyte fuel cell, microbial fuel cell, polymer electrolyte fuel cell and phosphoric acid fuel cells. It is used as a catalyst in hydrogenation reactions and a gas adsorbent. It can be coated on electrode surface by electroplating.