Cerium oxide, 20% in H2O, colloidal dispersion
Cerium oxide, 20% in H<sub>2</sub>O, colloidal dispersion
Cerium oxide, 20% in H<sub>2</sub>O, colloidal dispersion
Cerium oxide, 20% in H<sub>2</sub>O, colloidal dispersion
Thermo Scientific Chemicals
Cerium oxide, 20% in H2O, colloidal dispersion
CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Catalog NumberQuantity
ALF012730.36500 g
Catalog number ALF012730.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium oxide
Name NoteColloidal dispersion
CAS1306-38-3
ColorYellow
Health Hazard 1H290-H315-H319
View more
Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.