Selenous acid, 97%
Selenous acid, 97%
Selenous acid, 97%
Thermo Scientific Chemicals
Selenous acid, 97%
For Use As An Oxidizing Agent, As a Reagent For Alkaloids | CAS: 7783-00-8 | H2O3Se | 128.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF012574.065 g
Catalog number ALF012574.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSelenous acid
ColorColorless
CAS7783-00-8
Melting Point70°C (decomposition)
Health Hazard 1H301+H331-H373
View more
As an oxidizing agent, as a reagent for alkaloidsSelenous acid is used in steel guns, wherein it changes the color of the steel from silver-grey to blue-grey. It serves as an important component of the Mecke reagent, which is utilized for drug testing. It acts as a source of selenium. It acts as a reagent for alkaloids and an oxidizing agent. Further, it is used in labeling radiopharmaceuticals. It is involved in the organic synthesis for the preparation of glyoxal.