Chromium(III) 2,4-pentanedionate, 97%
Chromium(III) 2,4-pentanedionate, 97%
Chromium(III) 2,4-pentanedionate, 97%
Chromium(III) 2,4-pentanedionate, 97%
Thermo Scientific Chemicals
Chromium(III) 2,4-pentanedionate, 97%
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD | CAS: 21679-31-2 | C15H21CrO6 | 349.32 g/mol
Have Questions?
Catalog NumberQuantity
ALF012538.22100 g
Catalog number ALF012538.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialChromium(III) 2,4-pentanedionate
CAS21679-31-2
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVDChromium(III) 2,4-pentanedionate is used in the reduction of detonation of nitromethane. It is also used in NMR spectroscopy as a relaxation agent due to its solubility in nonpolar organic solvents and its paramagnetism. Further, it is involved in the modification of the surface properties of solid polyurethanes.