Iron(III) 2,4-pentanedionate
Iron(III) 2,4-pentanedionate
Iron(III) 2,4-pentanedionate
Iron(III) 2,4-pentanedionate
Thermo Scientific Chemicals
Iron(III) 2,4-pentanedionate
CAS: 14024-18-1 | C15H21FeO6 | 353.17 g/mol
Have Questions?
Catalog NumberQuantity
ALF012534.22100 g
Catalog number ALF012534.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIron(III) 2,4-pentanedionate
CAS14024-18-1
Health Hazard 1H302-H319-H335-H500
Health Hazard 2GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
View more
Iron(III) 2,4-pentanedionate is used as a precatalyst and reagent in organic chemistry. It acts as a precursor for highly crystalline (Zn, Fe)Fe2O4 films and magnetic property measurements of these films. It catalyzes the dimerization of isoprene to a mixture of 1,5-dimethyl-1,5-cyclooctadiene and 2,5-dimethyl-1,5-cyclooctadiene, ring-opening polymerization of 1,3-benzoxazine and catalyze the reaction of N-sulfonyl oxaziridines with olefins to form 1,3-oxazolidine products.