Nickel ammonium sulfate hexahydrate
Nickel ammonium sulfate hexahydrate
Nickel ammonium sulfate hexahydrate
Nickel ammonium sulfate hexahydrate
Thermo Scientific Chemicals
Nickel ammonium sulfate hexahydrate
CAS: 7785-20-8 | Ni(NH4)2(SO4)2·6H2O
Have Questions?
Catalog NumberQuantity
ALF012519.36500 g
Catalog number ALF012519.36
Price (MYR)
2,483.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,483.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel ammonium sulfate hexahydrate
CAS7785-20-8
ColorGreen
Health Hazard 1H302+H332-H317-H334-H341-H350i-H360D-H372
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
View more
It is used in electroplating. Has limited use as dye mordants and is used in metal-finishing compositions. Nickel electrolyte for electroplating