Iron(III) chloride hexahydrate, ACS, 97.0-102.0%
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%
Thermo Scientific Chemicals
Iron(III) chloride hexahydrate, ACS, 97.0-102.0%
Used to prepare organically templated iron phosphates.Also, used for sewerage treatment of industrial wastes, chloride hydrometallurgy. Useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids. | CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Catalog number ALF012497.A1
Price (MYR)
992.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
992.00
EA
Add to cart
Specifications
Boiling Point280°C to 285°C
Chemical Name or MaterialIron(III) Chloride Hexahydrate
Melting Point37°C
CAS10025-77-1
ColorBrown-Yellow
View more
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications:
  • Used to prepare organically templated Iron Phosphates
  • Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
  • Plays important role in photoengraving and photography as an etching agent
  • Usedl as mordant in dyeing and printing textiles.
  • Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids
RUO – Research Use Only