Strontium oxide, tech.
Strontium oxide, tech.
Strontium oxide, tech.
Strontium oxide, tech.
Thermo Scientific Chemicals
Strontium oxide, tech.
CAS: 1314-11-0 | OSr | 103.619 g/mol
Have Questions?
Catalog NumberQuantity
ALF012495.A11 kg
Catalog number ALF012495.A1
Price (MYR)
2,450.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
2,450.00
EA
Add to cart
Specifications
Boiling Point∼3000°C
Chemical Name or MaterialStrontium oxide
Melting Point2430°C
CAS1314-11-0
Health Hazard 1H314-H335
View more
In the manufacture of strontium salts, pyrotechnics, pigments, in greases and soaps and as a desiccant.Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industries. It is an active component of paint dryers. It is also used to prepare pure strontium upon heating with aluminum under vacuum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • In the manufacture of strontium salts, pyrotechnics, and pigments, in greases and soaps, and as a desiccant
RUO – Research Use Only