Bismuth Tin eutectic lump, 99.95% (metals basis)
Bismuth Tin eutectic lump, 99.95% (metals basis)
Thermo Scientific Chemicals
Bismuth Tin eutectic lump, 99.95% (metals basis)
Have Questions?
Catalog NumberQuantity
ALF012480.36500 g
Catalog number ALF012480.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth Tin eutectic lump
Melting Point138°C
Recommended StorageAmbient temperatures
Avg. Mol. Wt. or Mol. Wt. RangeBi:Sn; 58:42 wt%
TSCAYes