Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO)
Thermo Scientific Chemicals
Yttrium foil, 0.025mm (0.001in) thick, 99.9% (REO)
CAS: 7440-65-5 | Y | 88.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF012443.FI
Price (MYR)
2,789.00
EA
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
2,789.00
EA
Add to cart
Specifications
Chemical Name or MaterialYttrium foil
Name Note0.025mm (0.001 in.) thick
CAS7440-65-5
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228
Flammable solid.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only