Scandium foil, 0.025mm (0.001in) thick, 99% (REO)
Scandium foil, 0.025mm (0.001in) thick, 99% (REO)
Scandium foil, 0.025mm (0.001in) thick, 99% (REO)
Scandium foil, 0.025mm (0.001in) thick, 99% (REO)
Thermo Scientific Chemicals
Scandium foil, 0.025mm (0.001in) thick, 99% (REO)
CAS: 7440-20-2 | Sc | 44.96 g/mol
Have Questions?
Catalog NumberQuantity
ALF012426.FF25 x 25 mm
Catalog number ALF012426.FF
Price (MYR)
3,672.00
EA
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (MYR)
3,672.00
EA
Add to cart
Specifications
Chemical Name or MaterialScandium foil
ColorGray
Name Note0.025mm (0.001 in.) thick
CAS7440-20-2
Health Hazard 1H228
View more
It finds applications in the aerospace industry. Another use of Scandium is in aluminum-scandium alloys for sports equipment, such as bicycle frames, lacrosse sticks, and baseball bats.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only