Aluminum hydroxide, 76.5% min
Aluminum hydroxide, 76.5% min
Aluminum hydroxide, 76.5% min
Aluminum hydroxide, 76.5% min
Thermo Scientific Chemicals
Aluminum hydroxide, 76.5% min
CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF012366.22100 g
Catalog number ALF012366.22
Price (MYR)
233.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
233.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum hydroxide
CAS21645-51-2
Health Hazard 1H315-H319-H335
Recommended StorageAmbient temperatures
Physical FormGranules
View more
As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmeticsAluminum hydroxide is used as a desiccant powder, filler in paper, plastics, rubber and cosmetics. It is used as a smoke suppressant and mordant dye. It is also used in drugs as antacid and antihyperphosphatemic. It is also used as a Claus catalyst support for waterproof fabrics. It is an important starting material for the preparation of other aluminum compounds, calcined alumina, aluminum sulfate, polyaluminum chloride, zeolites, sodium aluminate, activated alumina and aluminum nitrate. In addition, it is used as a fire retardant.