Aluminum sulfate hydrate, 97+%
Aluminum sulfate hydrate, 97+%
Aluminum sulfate hydrate, 97+%
Aluminum sulfate hydrate, 97+%
Thermo Scientific Chemicals
Aluminum sulfate hydrate, 97+%
CAS: 17927-65-0 | Al2O12S3 | 342.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number ALF012362.A1
Price (MYR)
489.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
489.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum sulfate hydrate
Name Notehydrate
CAS17927-65-0
ColorWhite
Health Hazard 1H318
View more
Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.