Aluminum nitrate nonahydrate, 98%
Aluminum nitrate nonahydrate, 98%
Aluminum nitrate nonahydrate, 98%
Aluminum nitrate nonahydrate, 98%
Thermo Scientific Chemicals
Aluminum nitrate nonahydrate, 98%
CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Catalog number ALF012360.A1
Price (MYR)
881.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
881.00
EA
Add to cart
Specifications
Chemical Name or MaterialAluminum nitrate nonahydrate
Melting Point73.5°C (135° (decomposition))
Name Notenonahydrate
CAS7784-27-2
ColorWhite
View more
It finds use in tanning leather, antiperspirant, corrosion protection, extraction of uranium, nitrating agent. It is an effective dopant for producing highly conductive thin film by utilizing the sol gel method.