Cadmium sulfate octahydrate, ACS, 98.0-102.0%
Cadmium sulfate octahydrate, ACS, 98.0-102.0%
Cadmium sulfate octahydrate, ACS, 98.0-102.0%
Cadmium sulfate octahydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals
Cadmium sulfate octahydrate, ACS, 98.0-102.0%
Used in Standard Weston Cell, as electrolytes in electroplating, in phosphors, in manufacture of standard cadmium elements, and as a catalyst in Marsh test for AS | CAS: 7790-84-3 | 3CdSO4·8H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Catalog number ALF012340.30
Price (MYR)
728.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
728.00
EA
Add to cart
Specifications
Chemical Name or MaterialCadmium sulfate octahydrate
Melting Point>40°C (in -H2O)
CAS7790-84-3
ColorWhite
Health Hazard 1H301-H330-H340-H350-H360FD-H372
View more
Cadmium sulfate mainly used in electroplating of cadmium, which is used in electronic circuits. It is used as a precursor for cadmium based pigments such as cadmium sulfide and pharmaceutical intermediates. It is used in the standard weston cell.