Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min
Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min
Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min
Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min
Thermo Scientific Chemicals
Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min
CAS: 16940-92-4 | Cl6H8IrN2 | 440.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF012326.031 g
Catalog number ALF012326.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium hexachloroiridate(IV)
Melting Point(decomposition)
CAS16940-92-4
ColorBlack-Red
Health Hazard 1H302
View more
Iridium complexes are generated from ammonium hexachloroiridate.