Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min
Thermo Scientific Chemicals
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min
Used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments. | CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Catalog NumberQuantity
ALF012325.031 g
Catalog number ALF012325.03
Price (MYR)
1,107.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,107.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrogen tetrachloroaurate(III) hydrate
CAS27988-77-8
Health Hazard 1H302-H314-H317-H335
Health Hazard 3P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
Recommended StorageAmbient temperatures; Store under Nitrogen
View more
Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.