Calcium chloride, anhydrous, 93%
Calcium chloride, anhydrous, 93%
Calcium chloride, anhydrous, 93%
Calcium chloride, anhydrous, 93%
Thermo Scientific Chemicals
Calcium chloride, anhydrous, 93%
CAS: 10043-52-4 | CaCl2 | 110.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF012316.A11 kg
Catalog number ALF012316.A1
Price (MYR)
485.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
485.00
EA
Add to cart
Specifications
Boiling Point>1600°C
Chemical Name or MaterialCalcium chloride
Melting Point782°C
CAS10043-52-4
ColorWhite to Gray
View more
It is used as a drying and dehydrating agent for organic liquids and gases, and for solids in desiccators. Common applications include brine for refrigeration plants, ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranate. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishders, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium, and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.