Magnesium chloride, anhydrous, 99%
Magnesium chloride, anhydrous, 99%
Magnesium chloride, anhydrous, 99%
Magnesium chloride, anhydrous, 99%
Magnesium chloride, anhydrous, 99%
Thermo Scientific Chemicals
Magnesium chloride, anhydrous, 99%
For Use As a Catalyst, Flocculating Agent, Various Magnesium Cements | CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
Have Questions?
Catalog NumberQuantity
ALF012315.A11 kg
Catalog number ALF012315.A1
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium Chloride, Anhydrous
Melting Point714°C
CAS7786-30-3
ColorWhite to Yellow
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Hygroscopic; May contain trace amount of Zr as black insoluble particles
RUO – Research Use Only