Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Calcium chloride dihydrate, 99% min
Thermo Scientific Chemicals
Calcium chloride dihydrate, 99% min
CAS: 10035-04-8 | CaCl2·2H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF012312.A3
Price (MYR)
915.00
EA
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (MYR)
915.00
EA
Add to cart
Specifications
Chemical Name or MaterialCalcium chloride dihydrate
CAS10035-04-8
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
Recommended StorageAmbient temperatures
View more
Calcium chloride dihydrate is widely used as an ingredient in ceramic slipware, cosmetic and food items. It acts as a pH buffer in swimming pool water. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. It is employed in the oil industry to increase the density of solids-free brines. It is used as a flux in the Davy process for the manufacture of sodium through the electrolysis of molten sodium chloride. It finds an important application as an additive in plastics, fire extinguishers, blast furnaces and in wastewater treatment as a drainage aid.