Ammonium pentaborate octahydrate, 99.0%
Ammonium pentaborate octahydrate, 99.0%
Thermo Scientific Chemicals
Ammonium pentaborate octahydrate, 99.0%
CAS: 12046-03-6 | (NH4)2B10O16·8H2O
Have Questions?
Catalog NumberQuantity
ALF012294.36500 g
Catalog number ALF012294.36
Price (MYR)
533.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
533.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium Pentaborate
CAS12046-03-6
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
Ammonium pentaborate octahydrate is used as a component of electrolyte solutions in electrolytic capacitors. It is also used as a flame retarder and in wood and textile industry. Further, it is used for brazing, welding, soldering fluxes and in some detergents.