Boron oxide, 97.5% min, typically 98.5%
Boron oxide, 97.5% min, typically 98.5%
Boron oxide, 97.5% min, typically 98.5%
Boron oxide, 97.5% min, typically 98.5%
Thermo Scientific Chemicals
Boron oxide, 97.5% min, typically 98.5%
CAS: 1303-86-2 | B2O3 | 69.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF012290.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Boiling Point1860°C
Chemical Name or MaterialBoron oxide
Melting Point∽450°C
CAS1303-86-2
Health Hazard 1H360FD
View more
It is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from Talc.