Magnesium oxide, 96%, light
Magnesium oxide, 96%, light
Magnesium oxide, 96%, light
Magnesium oxide, 96%, light
Thermo Scientific Chemicals
Magnesium oxide, 96%, light
Bulk density: 4-8 lb./cu.ft. | CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number ALF012287.A1
Price (MYR)
1,468.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,468.00
EA
Add to cart
Specifications
Boiling Point3600°C
Chemical Name or MaterialMagnesium Oxide, Light
Melting Point2852°C
CAS1309-48-4
ColorWhite
View more
Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, a protective coating in plasma displays, an oxide barrier in spin-tunneling devices, an optical material, a component of tubular construction in heating elements, and also in libraries for preserving books. Magnesium oxide microspheres used in enrichment method of polycyclic aromatic hydrocarbons.