Chromium(III) oxide, 98+%
Chromium(III) oxide, 98+%
Chromium(III) oxide, 98+%
Chromium(III) oxide, 98+%
Thermo Scientific Chemicals
Chromium(III) oxide, 98+%
CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol
Have Questions?
Catalog NumberQuantity
ALF012286.36500 g
Catalog number ALF012286.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point3000°C
Chemical Name or MaterialChromium(III) oxide
Melting Point2435°C
CAS1308-38-9
ColorGreen
View more
It is employed in metallurgical processing and has applications in chrome plating, anodizing aluminum etc… It is also used as a pigment.