Mercury(II) chloride, ACS, 99.5% min
Mercury(II) chloride, ACS, 99.5% min
Mercury(II) chloride, ACS, 99.5% min
Mercury(II) chloride, ACS, 99.5% min
Mercury(II) chloride, ACS, 99.5% min
CAS: 7487-94-7 | Cl2Hg | 271.49 g/mol
Have Questions?
Catalog NumberQuantity
ALF012274.1425 g
Catalog number ALF012274.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point302°C
Chemical Name or MaterialMercury(II) chloride
Melting Point280°C to 282°C
CAS7487-94-7
ColorWhite
View more
Mercury(II) chloride is used as a catalyst in making plastics, depolarizer in batteries and as a reagent in organic synthesis and analytical chemistry. It is also used to prepare an amalgam with metals such as aluminum, which finds application as reducing agent in organic synthesis. It also finds application in photography as a photographic intensifier. Further, it serves as a preservative and used in the treatment of syphilis. It is also used to remove dithiane groups bonded to a carbonyl in an umpolung reaction.