Vanadium powder, -325 mesh, 99.5% (metals basis)
Vanadium powder, -325 mesh, 99.5% (metals basis)
Vanadium powder, -325 mesh, 99.5% (metals basis)
Vanadium powder, -325 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals
Vanadium powder, -325 mesh, 99.5% (metals basis)
CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 g
10 g
50 g
Catalog number ALF012234.30
Price (MYR)
9,615.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
9,615.00
EA
Add to cart
Specifications
Chemical Name or MaterialVanadium
ColorSilver to Gray
CAS7440-62-2
Melting Point1900°C
Boiling Point3000°C