Ammonium molybdate (di), Mo 56.5%
Ammonium molybdate (di), Mo 56.5%
Ammonium molybdate (di), Mo 56.5%
Ammonium molybdate (di), Mo 56.5%
Thermo Scientific Chemicals
Ammonium molybdate (di), Mo 56.5%
CAS: 27546-07-2 | H2MoO4 | 161.96 g/mol
Have Questions?
Catalog NumberQuantity
ALF012215.30250 g
Catalog number ALF012215.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium molybdate (di)
CAS27546-07-2
Recommended StorageAmbient temperatures
Physical FormPowder
Quantity250 g
View more
Ammonium molybdate (di) is used as pigments, agricultural fertilizer, color lakes and fabric fire retardant agent. It is also used as a raw material for the production of molybdenum wire, ceramic glazes and other molybdenum compounds. Further, it is used in photography, ceramics and paints. It is also used reagent for analysis as well as in the determination of phosphorus in analytical chemistry. In addition, it is used as a catalyst in the petrochemical industry.