Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
Thermo Scientific Chemicals
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
CAS: 7440-42-8 | B | 10.81 g/mol
Have Questions?
Catalog NumberQuantity
ALF012134.1850 g
Catalog number ALF012134.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBoron pieces
CAS7440-42-8
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.