Silver foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis)
Silver foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis)
Silver foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis)
Silver foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis)
Thermo Scientific Chemicals
Silver foil, 0.5mm (0.02in) thick, Premion™, 99.9985% (metals basis)
Form: Approx. 3.3g/25 x 25mm | CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF012128.GH100 x 100 mm
Catalog number ALF012128.GH
Price (MYR)
3,097.00
EA
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
3,097.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
CAS7440-22-4
AppearanceSilver-colored foil