Palladium powder, -200 mesh, 99.95% (metals basis)
Palladium powder, -200 mesh, 99.95% (metals basis)
Palladium powder, -200 mesh, 99.95% (metals basis)
Palladium powder, -200 mesh, 99.95% (metals basis)
Thermo Scientific Chemicals
Palladium powder, -200 mesh, 99.95% (metals basis)
CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Catalog NumberQuantity
ALF012068.031 g
Catalog number ALF012068.03
Price (MYR)
2,110.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
2,110.00
EA
Add to cart
Specifications
Chemical Name or MaterialPalladium
ColorGray to Black
CAS7440-05-3
Health Hazard 1H228-H315-H319-H335
AppearanceDark gray to black powder