Zinc foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)
Zinc foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)
Thermo Scientific Chemicals
Zinc foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Catalog NumberQuantity
ALF011913.FI50 x 50 mm
Catalog number ALF011913.FI
Price (MYR)
838.00
EA
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
838.00
EA
Add to cart
Specifications
Chemical Name or MaterialZinc
CAS7440-66-6
AppearanceGray foil
Density7.14g/cm3