Specifications
Adsorbent TypePolymer
Carbon Load0.86
Bed Weight200 mg
For Use With (Application)Extraction of strongly acidic compounds, highly stable in organic solvents
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size85 μm
Product LineSiliaPrepX
SorbentPolystyrene-divinylbenzene WAX
TypeSPE Cartridge
Volume (Metric)3 mL
MaterialPolypropylene (PP) tube, Polyethylene (PE) frits
Particle ShapeSpherical
Pore Size60 Å
Quantity50 Pk.
Surface Area (Metric)800 m2/g
Tube Size8.9 x 74.7 mm
Length (Metric)74.7 mm
Diameter (Metric) Inner8.9 mm
Unit SizeEA