SiliaPrepX™ SPE Cartridges - DVB
SiliaPrepX™ SPE Cartridges - DVB
Actual product may vary
Thermo Scientific Chemicals
SiliaPrepX™ SPE Cartridges - DVB
SPE Cartridges packed with polystyrene-divinylbenzene copolymer (DVB), for extraction of neutral, acidic or basic compounds.
Have Questions?
Catalog NumberBed WeightVolume (Metric)Quantity
ALF011829.18500 mg12 mL20 Pk.
Catalog number ALF011829.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Bed Weight:
500 mg
Volume (Metric):
12 mL
Quantity:
20 Pk.
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Adsorbent TypePolymer
Carbon Load9%
Bed Weight500 mg
For Use With (Application)Extraction of drugs and metabolites from biological fluids, API from tablets and cream
For Use With (Equipment)Vacuum Manifolds or Automated SPE Equipment
Particle Size85 μm
Product LineSiliaPrepX
SorbentDVB (Polystyrene-divinylbenzene)
TypeSPE Cartridge
Volume (Metric)12 mL
MaterialPolypropylene (PP) tube, Polyethylene (PE) frits
Particle ShapeSpherical
Pore Size60 Å
Quantity20 Pk.
Surface Area (Metric)1000 m2/g
Tube Size15.8 x 90 mm
Length (Metric)90 mm
Diameter (Metric) Inner15.8 mm
Unit SizeEA