E-PAK™ Cartridges, SiliaMetS™ DMT
E-PAK™ Cartridges, SiliaMetS™ DMT
Actual product may vary
Thermo Scientific Chemicals
E-PAK™ Cartridges, SiliaMetS™ DMT
E-PAK™ Cartridges, SiliaMetS™ DMT (2,4,6-trimercaptotriazine) is a silica-bound scavenger for a variety of metals, in particular, ruthenium catalysts and hindered Pd complexes conveniently packaged in a cartridge. E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
Have Questions?
Catalog NumberDimensions (L x W)Quantity
ALF011821.1810 x 5 cm10 Pk.
Catalog number ALF011821.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
10 x 5 cm
Quantity:
10 Pk.
Metals removed:
  • Best scavenger: Ir, Ni, Os, Pd, Pt, Rh, and Ru
  • Good scavenger: Cd, Co, Cu, Fe, Sc, and Zn
Additional Information:
  • To facilitate the evaluations of small samples, we can provide loose media and lab scale cartridges on a custom basis.
  • Testing with loose media can be done with samples as small as a few milliliters and are normally done in advance of cartridge testing to identify the formula with the highest capacity to remove contaminants with the least loss of product.
  • E-PAK™ lab cartridges are generally used for processing samples of 50 mL or more.
  • Testing has shown good correlation between the performance of lab, pilot and commercial size cartridges.
  • Lab cartridges can be operated with pump or small pressure vessel.
Specifications
TypeLab Scale E-PAK Cartridge
For Use With (Application)Lower residual metal concentration in Ir, Ni, Os, Pd, Pt, Rh and Ru
ColorWhite to yellow
Dimensions (L x W)10 x 5 cm
Quantity10 Pk.
pH2 to 9
Particle Size40 to 63 μm
SorbentDMT
Sorbent Weight75 g
Unit SizeEA